Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Methyl-2-(trimethylsilyl)pyridine ≥95%

Supplier: AOB CHEM USA

478030-5G 478030-1G 478030-25G 478030-500MG
77633-338EA 1187 USD InStock
77633-338 77639-166 77659-654 77651-166
5-Methyl-2-(trimethylsilyl)pyridine ≥95%
5-Methyl-2-(trimethylsilyl)pyridine
Precaución: For Research Use Only


Formula: 165.308 g/mol
CAS Number: 13737-09-2
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty