Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-(2,5-diethylphenyl)-2-oxoacetate ≥95%

Supplier: AOB CHEM USA

478117-5G 478117-25G 478117-1G 478117-500MG
77629-768EA 1484.23 USD InStock
77629-768 77635-946 77639-170 77647-204
Ethyl 2-(2,5-diethylphenyl)-2-oxoacetate ≥95%
Ethyl 2-(2,5-diethylphenyl)-2-oxoacetate
Precaución: For Research Use Only


Formula: 234.291 g/mol
CAS Number: 1407280-44-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty