Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tert-butyl 3-fluoro-2-methoxyisonicotinate ≥95%

Supplier: AOB CHEM USA

480551-5G 480551-25G 480551-1G 480551-500MG
77629-798EA 1532.64 USD InStock
77629-798 77635-954 77639-200 77651-190
Tert-butyl 3-fluoro-2-methoxyisonicotinate ≥95%
Tert-butyl 3-fluoro-2-methoxyisonicotinate
Precaución: For Research Use Only


Formula: 227.232 g/mol
CAS Number: 3031789-29-1
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty