Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Fluoro-3,5-dimethoxybenzaldehyde ≥97%

Supplier: AOB CHEM USA

49459-5G 49459-25G 49459-1G 49459-500MG
77633-444EA 1216 USD InStock
77633-444 77656-294 77643-440 77647-394
2-Fluoro-3,5-dimethoxybenzaldehyde ≥97%
2-Fluoro-3,5-dimethoxybenzaldehyde
Precaución: For Research Use Only


Formula: 184.164 g/mol
MDL Number: MFCD20482718
CAS Number: 113984-71-7
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty