Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(1H-Pyrazol-1-yl)isonicotinic acid ≥97%

Supplier: AOB CHEM USA
Warning

507822-5G 507822-25G 507822-1G 507822-500MG
77635-124EA 518 USD InStock
77635-124 77656-342 77643-502 77647-454
2-(1H-Pyrazol-1-yl)isonicotinic acid ≥97%
2-(1H-Pyrazol-1-yl)isonicotinic acid
Precaución: For Research Use Only


Formula: C₉H₇N₃O₂
Storage Temperature: Ambient
MDL Number: MFCD11128544
CAS Number: 1152940-72-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty