Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl [3,5-bis(trifluoromethyl)phenyl](oxo)acetate ≥95%

Supplier: AOB CHEM USA
Warning

82431-5G 82431-25G 82431-1G 82431-500MG
77633-742EA 1166 USD InStock
77633-742 77660-390 77644-048 77652-038
Ethyl [3,5-bis(trifluoromethyl)phenyl](oxo)acetate ≥95%
Ethyl [3,5-bis(trifluoromethyl)phenyl](oxo)acetate
Precaución: For Research Use Only


Formula: C₁₂H₈F₆O₃
Storage Temperature: Ambient
MDL Number: MFCD03094082
CAS Number: 402568-10-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty