Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,3,4-Trichloroanisole ≥95%

Supplier: AOB CHEM USA
Warning

87331-5G 87331-1G 87331-25G 87331-500MG
77633-816EA 1029 USD InStock
77633-816 77640-124 77660-472 77648-126
2,3,4-Trichloroanisole ≥95%
2,3,4-Trichloroanisole
Precaución: For Research Use Only


Formula: C₇H₅Cl₃O
Storage Temperature: Ambient
MDL Number: MFCD00000541
CAS Number: 54135-80-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty