Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-3-pyridinemethanol ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A119400-100G A119400-1G A119400-25G A119400-5G
ALA119400-100GEA 2210.56 USD InStock
ALA119400-100G ALA119400-1G ALA119400-25G ALA119400-5G
2-Amino-3-pyridinemethanol ≥98%
2-Amino-3-pyridinemethanol
A pyridine derivative for proteomics research


Formula: C₆H₈N₂O
Boiling Pt: 279 °C
Melting Pt: 65.0-70.0 °C
Storage Temperature: Ambient
MDL Number: MFCD05663510
CAS Number: 23612-57-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty