Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(1-Adamantyl)-4-bromoanisole ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A120005-100G A120005-25G A120005-5G
ALA120005-100GEA 1706.11 USD InStock
ALA120005-100G ALA120005-25G ALA120005-5G
2-(1-Adamantyl)-4-bromoanisole ≥98%
2-(1-Adamantyl)-4-bromoanisole


Formula: C₁₇H₂₁BrO
Melting Pt: 144°C
MDL Number: MFCD03855308
CAS Number: 104224-63-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty