Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A120024-100G A120024-1G A120024-200MG A120024-25G A120024-5G
ALA120024-100GEA 2997.16 USD InStock
ALA120024-100G ALA120024-1G ALA120024200MG ALA120024-25G ALA120024-5G
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate ≥97%
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate


Formula: C₁₄H₁₉BO₄
Storage Temperature: Ambient
MDL Number: MFCD03453060
CAS Number: 480424-70-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty