Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1'-(Azodicarbonyl)dipiperidine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A120043-10G A120043-1G A120043-25G A120043-5G
ALA120043-10GEA 127.21 USD InStock
ALA120043-10G ALA120043-1G ALA120043-25G ALA120043-5G
1,1'-(Azodicarbonyl)dipiperidine ≥98%
1,1'-(Azodicarbonyl)dipiperidine
Used in a study of the copper-catalyzed addition of aryboronic acids to azodicarboxyl derivatives providing aryl-substituted hydrazides. Reactant for: Mitsunobu inversion reactions


Formula: C₁₂H₂₀N₄O₂
MW: 252.32 g/mol
Melting Pt: 133-136°C
MDL Number: MFCD00010111
CAS Number: 10465-81-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty