Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-3-bromo-2-chloropyridine ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

A120067-100G A120067-1G A120067-25G A120067-5G
ALA120067-100GEA 658.26 USD InStock
ALA120067-100G ALA120067-1G ALA120067-25G ALA120067-5G
5-Amino-3-bromo-2-chloropyridine ≥97%
5-Amino-3-bromo-2-chloropyridine


Formula: C₅H₄BrClN₂
MW: 207.46
Melting Pt: 118-123°C
MDL Number: MFCD03840433
CAS Number: 130284-53-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty