Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-4-methylphenylboronic acid ≥98% (contains varying amounts of Anhydride)

Supplier: ALADDIN SCIENTIFIC
Warning

A120097-1G A120097-25G A120097-5G
ALA120097-1GEA 55.96 USD InStock
ALA120097-1G ALA120097-25G ALA120097-5G
3-Amino-4-methylphenylboronic acid ≥98% (contains varying amounts of Anhydride)
3-Amino-4-methylphenylboronic acid


Formula: C₇H₁₀BNO₂
MW: 150.97 g/mol
Melting Pt: 250°C
MDL Number: MFCD01074640
CAS Number: 22237-12-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty