Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-pyrimidineboronic acid pinacol ester ≥96%

Supplier: ALADDIN SCIENTIFIC
Danger

A120137-1G A120137-250MG A120137-5G
ALA120137-1GEA 77.81 USD InStock
ALA120137-1G ALA120137250MG ALA120137-5G
2-Amino-5-pyrimidineboronic acid pinacol ester ≥96%
2-Amino-5-pyrimidineboronic acid pinacol ester


Formula: C₁₀H₁₆BN₃O₂
MW: 221.06 g/mol
Melting Pt: 212 °C
MDL Number: MFCD06795685
CAS Number: 402960-38-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty