Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2-fluorobenzoic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A120664-250MG A120664-25G A120664-5G A120664-100G A120664-1G
ALA120664250MGEA 9.41 USD InStock
ALA120664250MG ALA120664-25G ALA120664-5G ALA120664-100G ALA120664-1G
4-Amino-2-fluorobenzoic acid ≥98%
4-Amino-2-fluorobenzoic acid


Formula: C₇H₆FNO₂
Melting Pt: 209-211°C
Storage Temperature: Ambient
MDL Number: MFCD01569397
CAS Number: 446-31-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty