Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Bromoanthranilic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A120954-100G A120954-1G A120954-25G A120954-5G
ALA120954-100GEA 539.51 USD InStock
ALA120954-100G ALA120954-1G ALA120954-25G ALA120954-5G
4-Bromoanthranilic acid ≥98%
4-Bromoanthranilic acid


Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 230-234°C
MDL Number: MFCD03618454
CAS Number: 20776-50-5
EINECS: 244-025-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty