Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-bromo-3-methoxypyrazine ≥95%

Supplier: ALADDIN SCIENTIFIC
Danger

A121884-500MG A121884-5G A121884-1G
ALA121884500MGEA 43.61 USD InStock
ALA121884500MG ALA121884-5G ALA121884-1G
2-Amino-5-bromo-3-methoxypyrazine ≥95%
2-Amino-5-bromo-3-methoxypyrazine


Formula: C₅H₆BrN₃O
Melting Pt: 135-140°C
Storage Temperature: Refrigerator
MDL Number: MFCD08275726
CAS Number: 5900-13-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty