Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Amino-3,6,9,12,15,18,21,24-octaoxaheptacosan-27-oic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A122172-100MG A122172-1G A122172-250MG A122172-5G
ALA122172100MGEA 150.01 USD InStock
ALA122172100MG ALA122172-1G ALA122172250MG ALA122172-5G
1-Amino-3,6,9,12,15,18,21,24-octaoxaheptacosan-27-oic acid ≥98%
1-Amino-3,6,9,12,15,18,21,24-octaoxaheptacosan-27-oic acid


Formula: C₁₉H₃₉NO₁₀
Storage Temperature: Freezer
MDL Number: MFCD11041146
CAS Number: 756526-04-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty