Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A122371-1G A122371-25G A122371-5G
ALA122371-1GEA 39.81 USD InStock
ALA122371-1G ALA122371-25G ALA122371-5G
(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid ≥98%
(S)-4-Amino-2-(tert-butoxycarbonylamino)butyric acid


Formula: C₉H₁₈N₂O₄
MW: 218.25 g/mol
Melting Pt: 202 °C
MDL Number: MFCD00236841
CAS Number: 25691-37-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty