Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2-chloropyrimidine-5-carbonitrile ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A123179-1G A123179-25G A123179-500MG A123179-5G
ALA123179-1GEA 143.36 USD InStock
ALA123179-1G ALA123179-25G ALA123179500MG ALA123179-5G
4-Amino-2-chloropyrimidine-5-carbonitrile ≥97%
4-Amino-2-chloropyrimidine-5-carbonitrile


Formula: C₅H₃ClN₄
Melting Pt: 243-245°C
MDL Number: MFCD00052343
CAS Number: 94741-69-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty