Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Methoxyanthranilic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A123216-25G A123216-5G A123216-100G A123216-1G
ALA123216-25GEA 6213.86 USD InStock
ALA123216-25G ALA123216-5G ALA123216-100G ALA123216-1G
6-Methoxyanthranilic acid ≥97%
6-Methoxyanthranilic acid


Formula: C₈H₉NO₃
MW: 167.16
Melting Pt: 85°C
MDL Number: MFCD01941328
CAS Number: 53600-33-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty