Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminoacetophenone hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A123237-100G A123237-25G A123237-5G
ALA123237-100GEA 844.46 USD InStock
ALA123237-100G ALA123237-25G ALA123237-5G
2-Aminoacetophenone hydrochloride ≥98%
2-Aminoacetophenone hydrochloride


Formula: C₈H₁₀ClNO
Melting Pt: 194°C
MDL Number: MFCD00012873
CAS Number: 5468-37-1
EINECS: 226-787-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty