Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2-chloropyrimidine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A123454-100G A123454-1G A123454-250MG A123454-25G A123454-5G
ALA123454-100GEA 2428.11 USD InStock
ALA123454-100G ALA123454-1G ALA123454250MG ALA123454-25G ALA123454-5G
4-Amino-2-chloropyrimidine ≥98%
4-Amino-2-chloropyrimidine


Formula: C₄H₄ClN₃
MW: 129.55
Melting Pt: 197-205°C
MDL Number: MFCD00194051
CAS Number: 7461-50-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty