Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Aminopyridazine ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Warning

A123467-1G A123467-5G
ALA123467-1GEA 19.86 USD InStock
ALA123467-1G ALA123467-5G
3-Aminopyridazine ≥98% (by GC)
3-Aminopyridazine


Formula: C4H5N3
MW: 95.10 g/mol
Melting Pt: 171°C
Storage Temperature: Ambient
MDL Number: MFCD01529869
CAS Number: 5469-70-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty