Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromoanthranilic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

A123772-100G A123772-1G A123772-25G A123772-5G
ALA123772-100GEA 1947.41 USD InStock
ALA123772-100G ALA123772-1G ALA123772-25G ALA123772-5G
3-Bromoanthranilic acid ≥97%
3-Bromoanthranilic acid
Used as pharmaceutical intermediates.


Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 174-178°C
Flash Pt: 161 °C
MDL Number: MFCD03618453
CAS Number: 20776-51-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty