Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Bromoanthranilic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A123860-100G A123860-10G A123860-1G
ALA123860-100GEA 5198.31 USD InStock
ALA123860-100G ALA123860-10G ALA123860-1G
6-Bromoanthranilic acid ≥98%
6-Bromoanthranilic acid


Formula: C₇H₆BrNO₂
MDL Number: MFCD03618455
CAS Number: 20776-48-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty