Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-2-chlorobenzoic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A123909-100G A123909-25G A123909-500G A123909-5G
ALA123909-100GEA 510.06 USD InStock
ALA123909-100G ALA123909-25G ALA123909-500G ALA123909-5G
4-Amino-2-chlorobenzoic acid ≥98%
4-Amino-2-chlorobenzoic acid


Formula: C₇H₆ClNO₂
Melting Pt: 206.0-216.0°C
Storage Temperature: Ambient
MDL Number: MFCD00007772
CAS Number: 2457-76-3
EINECS: 219-540-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty