Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-3-azabicyclo[3.3.0]octane hydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A124225-100G A124225-25G A124225-5G
ALA124225-100GEA 1202.61 USD InStock
ALA124225-100G ALA124225-25G ALA124225-5G
3-Amino-3-azabicyclo[3.3.0]octane hydrochloride ≥97%
3-Amino-3-azabicyclo[3.3.0]octane hydrochloride
3-Amino-3-azabicyclo[3.3.0]octane hydrochloride is also referred to as N-amino-3-azabicyclo[3.3.0]octane monohydrochloride.


Formula: C₇H₁₅ClN₂
Melting Pt: 170-174°C
MDL Number: MFCD00792451
CAS Number: 58108-05-7
EINECS: 261-131-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty