Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-nitropyridine ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A124302-100G A124302-25G
ALA124302-100GEA 207.96 USD InStock
ALA124302-100G ALA124302-25G
2-Amino-5-nitropyridine ≥98%
2-Amino-5-nitropyridine


Formula: C₅H₅N₃O₂
Melting Pt: 185-190°C
Flash Pt: 224°C
Storage Temperature: Ambient
MDL Number: MFCD00006325
CAS Number: 4214-76-0
EINECS: 224-145-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty