Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Aprepitant ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

A125005-100MG A125005-1G A125005-250MG A125005-25G A125005-5G
ALA125005100MGEA 122.46 USD InStock
ALA125005100MG ALA125005-1G ALA125005250MG ALA125005-25G ALA125005-5G
Aprepitant ≥98%, Moligand™
Aprepitant
Aprepitant is a potent and selective neurokinin-1 receptor antagonist


Formula: C₂₃H₂₁F₇N₄O₃
Melting Pt: 252 °C
Storage Temperature: Freezer
MDL Number: MFCD08277635
CAS Number: 170729-80-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty