Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AZD-5438 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC

A126894-10MG A126894-5MG
ALA126894-10MGEA 94.91 USD InStock
ALA126894-10MG ALA126894-5MG
AZD-5438 ≥98%, Moligand™
AZD-5438


Formula: C₁₈H₂₁N₅OS
MW: 371.46 g/mol
Storage Temperature: Freezer
MDL Number: MFCD11112135
CAS Number: 602306-29-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty