Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AZD-7762 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Danger

A127862-100MG A127862-1MG A127862-25MG A127862-5MG
ALA127862100MGEA 535.71 USD InStock
ALA127862100MG ALA127862-1MG ALA127862-25MG ALA127862-5MG
AZD-7762 ≥98%, Moligand™
AZD-7762
AZD7762 is a potent and selective inhibitor of Chk1 with IC50 of 5 nM in a cell-free assay. It is equally potent against Chk2 and less potent against CAM, Yes, Fyn, Lyn, Hck and Lck.A checkpoint kinase inhibitor for Chk1 and Chk2


Formula: C₁₇H₁₉FN₄O₂S
Storage Temperature: Freezer
MDL Number: MFCD12406007
CAS Number: 860352-01-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty