Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

A66 ≥98%

Supplier: ALADDIN SCIENTIFIC

A128051-50MG A128051-5MG A128051-10MG A128051-1MG A128051-25MG
ALA128051-50MGEA 507.21 USD InStock
ALA128051-50MG ALA128051-5MG ALA128051-10MG ALA128051-1MG ALA128051-25MG
A66 ≥98%
A66


Formula: C₁₇H₂₃N₅O₂S₂
Storage Temperature: Freezer
MDL Number: MFCD22378485
CAS Number: 1166227-08-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty