Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Aminonicotinic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A128112-100G A128112-1G A128112-25G A128112-5G
ALA128112-100GEA 354.26 USD InStock
ALA128112-100G ALA128112-1G ALA128112-25G ALA128112-5G
5-Aminonicotinic acid ≥98%
5-Aminonicotinic acid


Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: 293°C
Storage Temperature: Ambient
MDL Number: MFCD00129116
CAS Number: 24242-19-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty