Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AZ-960 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC

A129464-50MG A129464-5MG A129464-100MG A129464-10MG A129464-25MG A129464-2MG
ALA129464-50MGEA 1487.61 USD InStock
ALA129464-50MG ALA129464-5MG ALA129464100MG ALA129464-10MG ALA129464-25MG ALA129464-2MG
AZ-960 ≥98%, Moligand™
AZ-960


Formula: C₁₈H₁₆F₂N₆
MW: 354.36 g/mol
Storage Temperature: Freezer
MDL Number: MFCD16621127
CAS Number: 905586-69-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty