Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

AZD-1981 ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

A129498-25MG A129498-5MG
ALA129498-25MGEA 209.86 USD InStock
ALA129498-25MG ALA129498-5MG
AZD-1981 ≥98%, Moligand™
AZD-1981
AZD1981 is a potent, selective CRTh2 (DP2) receptor antagonist with IC50 of 4 nM, showing >1000-fold selectivity over more than 340 other enzymes and receptors, including DP1.


Formula: C₁₉H₁₇ClN₂O₃S
MW: 388.87 g/mol
Storage Temperature: Freezer
MDL Number: MFCD26936340
CAS Number: 802904-66-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty