Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Fluorophenyl)-[(3S)-3-[3-(4-fluorophenyl)-1,2,4-oxadiazol-5-yl]piperidin-1-yl]methanone ≥98%, Moligand™

Supplier: ALADDIN SCIENTIFIC

A129508-10MG A129508-50MG A129508-5MG
ALA129508-10MGEA 207.01 USD InStock
ALA129508-10MG ALA129508-50MG ALA129508-5MG
(4-Fluorophenyl)-[(3S)-3-[3-(4-fluorophenyl)-1,2,4-oxadiazol-5-yl]piperidin-1-yl]methanone ≥98%, Moligand™
(4-Fluorophenyl)-[(3S)-3-[3-(4-fluorophenyl)-1,2,4-oxadiazol-5-yl]piperidin-1-yl]methanone


Formula: C₂₀H₁₇F₂N₃O₂
Storage Temperature: Freezer
MDL Number: MFCD17167026
CAS Number: 851881-60-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty