Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-5-chlorobenzoxazole ≥99%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

A129619-25G A129619-5G A129619-1G
ALA129619-25GEA 978.41 USD InStock
ALA129619-25G ALA129619-5G ALA129619-1G
2-Amino-5-chlorobenzoxazole ≥99%, Moligand™
2-Amino-5-chlorobenzoxazole


Formula: C₇H₅ClN₂O
MW: 168.58
Boiling Pt: 309
Melting Pt: 184
Storage Temperature: Freezer
MDL Number: MFCD00005770
CAS Number: 61-80-3
EINECS: 200-519-4
Merck Index: 12,10328
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty