Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Axitinib ≥99%, Moligand™

Supplier: ALADDIN SCIENTIFIC
Warning

A129732-1G A129732-250MG A129732-50MG
ALA129732-1GEA 197.51 USD InStock
ALA129732-1G ALA129732250MG ALA129732-50MG
Axitinib ≥99%, Moligand™
Axitinib
Axitinib is a non-selective receptor kinase inhibitor of Flt and Flk proteins. This compound also inhibits Flt-1(VEGFR-1), Flk-1(VEGFR-2), Flt-4(VEGFR-3), PDGFR-β and c-Kit(c-KIT) with IC50 of 0.1 nM, 0.2 nM, 0.1-0.3 nM, 1.6 nM and 1.7 nM, respectively. Axitinib is studied as an antitumor agent of various cell lines. Axitinib is an inhibitor of PDGFR-β.An inhibitor of Flt and Flk (VEGFR) with antitumor properties


Application:

Axitinib has been used as a vascular endothelial growth factor (VEGF) receptor tyrosine kinase inhibitor (TKI) to study its effect on renal cancer cell proliferation in vitro. It has also been used to inhibit anterior mesenteric artery (AMA) angiogenesis in dechlorinated embryos maintained in conditioned water (CW).


Formula: C₂₂H₁₈N₄OS
MW: 386.5 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09837898
CAS Number: 319460-85-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty