Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Aminoazotoluene ≥97% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Danger

A131595-100G A131595-25G
ALA131595-100GEA 65.46 USD InStock
ALA131595-100G ALA131595-25G
2-Aminoazotoluene ≥97% (by HPLC)
2-Aminoazotoluene
Fast Garnet GBC base has been used:· as a stain in papain activity inhibition assay.· to stain for tryptase-activity and chymase-like activity.· to gel stain for esterolytic activity.


Formula: C₁₄H₁₅N₃
Melting Pt: 101-102 °C
Density: 1.09 g/cm³
MDL Number: MFCD00007733
CAS Number: 97-56-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty