Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Amiloride hydrochloride ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A131615-1G A131615-5G
ALA131615-1GEA 164.26 USD InStock
ALA131615-1G ALA131615-5G
Amiloride hydrochloride ≥98%
Amiloride hydrochloride
Amiloride ? HCl is a selective T-type calcium channel protein inhibitor that has been shown to inhibit the Polycystin-L channel (Polycystic kidney disease 2-like 1 protein: PKD2L1), a member of the transient receptor potential (TRP) superfamily of cation channels, including the NHE (sodium/hydgrogen exchanger). Amiloride is also an epithelial sodium channel protein inhibitor, which inhibits sodium reabsorption without affecting potassium concentrations. Amiloride serves as a non-selective acid-sensing ion channel (ASIC) blocker and a selective inhibitor of uPA (urokinase plasminogen activator). Research shows that amiloride reduces electrical potential across tubular epithelium, inhibits angiogenesis and inhibits capillary morphogenesis completely and reversibly at approximately 130 μM. Amiloride ? HCl is an inhibitor of Sodium/Potassium-ATPase Protein.A calcium channel andsodium channel protein inhibitor


Formula: C₆H₈ClN₇O·HCl
Melting Pt: 299°C
Storage Temperature: Freezer
MDL Number: MFCD00058197
CAS Number: 2016-88-8
UN: 2811
ADR: 6.1,I
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty