Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α-Fluorophenylacetic acid ≥97%

Supplier: ALADDIN SCIENTIFIC
Danger

A131766-1G A131766-250MG A131766-5G
ALA131766-1GEA 89.21 USD InStock
ALA131766-1G ALA131766250MG ALA131766-5G
α-Fluorophenylacetic acid ≥97%
α-Fluorophenylacetic acid


Formula: C₈H₇FO₂
MW: 154.14 g/mol
Melting Pt: 78-82°C
MDL Number: MFCD00004221
CAS Number: 1578-63-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty