Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Sodium 4-acetamidobenzenesulfinate dihydrate ≥98% (by titration)

Supplier: ALADDIN SCIENTIFIC

A132494-1G A132494-25G A132494-5G
ALA132494-1GEA 44.56 USD InStock
ALA132494-1G ALA132494-25G ALA132494-5G
Sodium 4-acetamidobenzenesulfinate dihydrate ≥98% (by titration)
Sodium 4-acetamidobenzenesulfinate dihydrate


Formula: C₈H₉NO₃S
MW: 221.21 g/mol
MDL Number: MFCD04113633
CAS Number: 15898-43-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty