Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-(Aminomethyl)benzonitrile hydrochloride ≥98% (by HPLC)

Supplier: ALADDIN SCIENTIFIC
Warning

A132657-100G A132657-25G A132657-5G
ALA132657-100GEA 855.86 USD InStock
ALA132657-100G ALA132657-25G ALA132657-5G
4-(Aminomethyl)benzonitrile hydrochloride ≥98% (by HPLC)
4-(Aminomethyl)benzonitrile hydrochloride
4-(Aminomethyl)benzonitrile hydrochloride can be used in the synthesis of fluorescent-labeled bisbenzamidine, which can be a biochemical tool in biomedical studies. It can also be used in the synthesis of 3-(p-benzylamino)-1,2,4,5 tetrazine, used in the modification of alginate hydrogels, employed in cell engineering applications.


Formula: C₈H₉ClN₂
Melting Pt: 274-284°C
MDL Number: MFCD01861472
CAS Number: 15996-76-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty