Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Alamethicin ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A132913-25MG A132913-5MG
ALA132913-25MGEA 1592.11 USD InStock
ALA132913-25MG ALA132913-5MG
Alamethicin ≥98%
Alamethicin
Alamethicin (U-22324) is a polypeptide natural product antibiotic supplied as a mixture of homologs: Alamethicin and Alamethicin I. These compounds form voltage-dependent ion channels in the lipid bilayer, through the spontaneous assembly of units into a multimeric barrel-stave structure spanning the membrane. This formation results in a hydrophilic pore which permits non-specific ion efflux through the membrane. The non-specific ion efflux obliterates the normal electrochemical gradient of the cell, producing the antibiotic property of Alamethicin. Substantiated by experiments with covalently tethered molecules of Alamethicin, It is suggested that different multimeric aggregation states create varying pore sizes with varying stabilities, correlating to differential specificity for ions. This channel-forming activity has been exploited as a tool for introducing hydrophilic small molecules into mitochondria without disrupting the integrity of the membrane.Alamethicin is a monovalent cation ionophore which can mimic nerve action potential across artificial membranes. Alamethicin has also been used for studying membrane interactions of antimicrobial peptides.


Formula: C₉₂H₁₅₀N₂₂O₂₅
Melting Pt: 252°C
Storage Temperature: Freezer
MDL Number: MFCD00151517
CAS Number: 59588-86-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty