Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Amino-4,6-dichlorophenol ≥98% (by titration)

Supplier: ALADDIN SCIENTIFIC
Danger

A133472-1G A133472-25G A133472-5G A133472-100G A133472-10G
ALA133472-1GEA 66.41 USD InStock
ALA133472-1G ALA133472-25G ALA133472-5G ALA133472-100G ALA133472-10G
2-Amino-4,6-dichlorophenol ≥98% (by titration)
2-Amino-4,6-dichlorophenol


Formula: C₆H₅Cl₂NO
MDL Number: MFCD00035766
CAS Number: 527-62-8
UN: 2811
ADR: 6.1,I
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty