Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Allyl furan-2-carboxylate ≥98%

Supplier: ALADDIN SCIENTIFIC
Warning

A133703-100G A133703-25G A133703-5G
ALA133703-100GEA 220.31 USD InStock
ALA133703-100G ALA133703-25G ALA133703-5G
Allyl furan-2-carboxylate ≥98%
Allyl furan-2-carboxylate


Formula: C₈H₈O₃
Storage Temperature: Refrigerator
MDL Number: MFCD00016580
CAS Number: 4208-49-5
EINECS: 224-128-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty