Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-1H-indazole ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

A134463-100G A134463-1G A134463-250MG A134463-25G A134463-5G
ALA134463-100GEA 91236.01 USD InStock
ALA134463-100G ALA134463-1G ALA134463250MG ALA134463-25G ALA134463-5G
3-Amino-1H-indazole ≥97%
3-Amino-1H-indazole


Formula: C₇H₇N₃
Melting Pt: 150-153°C
Storage Temperature: Freezer
MDL Number: MFCD00182045
CAS Number: 874-05-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty