Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Amino-3-bromobenzoic acid ≥98%

Supplier: ALADDIN SCIENTIFIC
Danger

A134517-1G A134517-25G A134517-5G
ALA134517-1GEA 10.36 USD InStock
ALA134517-1G ALA134517-25G ALA134517-5G
4-Amino-3-bromobenzoic acid ≥98%
4-Amino-3-bromobenzoic acid


Formula: C₇H₆BrNO₂
Melting Pt: 211-215°C
MDL Number: MFCD03407439
CAS Number: 6311-37-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty