Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-2,5-dichloropyridine ≥98% (by GC)

Supplier: ALADDIN SCIENTIFIC
Danger

A136471-100G A136471-1G A136471-25G A136471-5G
ALA136471-100GEA 5131.81 USD InStock
ALA136471-100G ALA136471-1G ALA136471-25G ALA136471-5G
3-Amino-2,5-dichloropyridine ≥98% (by GC)
3-Amino-2,5-dichloropyridine


Formula: C₅H₄Cl₂N₂
Melting Pt: 127 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00204157
CAS Number: 78607-32-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty